Eisco Dropping Funnels with PTFE Stopcock, Open Top
Supplier: Wards
CH0470B CH0470E CH0470A CH0470D CH0470C
470343-858EA 60.45 CAD
470343-858 470344-320 470345-956 470345-998 470346-000
Eisco Dropping Funnels with PTFE Stopcock, Open Top
Borosilicate glass dropping funnels with PTFE stopcocks and open tops, ideal for basic extractions.
  • Cylindrical ungraduated glass dropping funnel
  • Made of durable, high quality borosilicate 3.3 glass which withstands high temperatures and thermal shocks
  • Open top for easily adding liquids
  • Fitted with a PTFE key stopcock
  • Used for extractions or to dispense liquids into a system slowly (drop-wise)
Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C