Eisco Anti-Fog Safety Goggles
Supplier: Wards
G104 G103
470344-420EA 7.89 CAD
470344-420 470345-818
Eisco Anti-Fog Safety Goggles
Goggles
Anti-fog safety goggles protect eyes from chemical splashes, high speed particles, dust and UV wavelengths (360-380).
  • Lightweight, durable polycarbonate lens
  • Soft, flexible vinyl frame and pliable nose bridge provides a comfortable and secure fit
  • Designed to be able to fit over prescription eyeglasses
  • Ventilated to prevent fogging of the goggle lenses
  • Features a freely adjustable, soft elastic head strap for a universal fit
Industrial quality safety goggles measure 6" wide, 3" long and 3" tall. Adjustable, head strap elastic measures 13.5" in total length. Direct ventilated goggles feature grids holes surrounding the entire frame, while indirect ventilated goggles feature four hooded vents for added splash protection.