Microscope Cover Glasses, Rectangular, No. 1
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
SCG218118 SCG318118 SCG222122 SCG322122 SCG224150 SCG324150
76970-088BX 6.75 CAD
76970-088 76970-090 76970-092 76970-094 76970-096 76970-098
Microscope Cover Glasses, Rectangular, No. 1
Cover Glasses
Four E's premium cover glasses with available selection between super white glass or borosilicate glass 3.3
  • Precision-cut for uniform sized pieces
  • Ultra-clear glass for optimal visibility
  • Premium quality material for durability
  • Autoclavable
No. 1 pertains to 0.13 to 0.16 mm thickness. The super white glass offering allows for superior light transmittance for enhanced visibility.