Silane-Coated Adhesive Microscope Slides, Color Frosted
Supplier: FOUR E#S INDUSTRIAL GROUP INC.
SLD245221 SLD245222 SLD245223 SLD245224
76970-080BX 39.75 CAD
76970-080 76970-082 76970-084 76970-086
Silane-Coated Adhesive Microscope Slides, Color Frosted
Slides Microscope Slides
Four E's premium adhesive glass slides with silane coating for superior adhesion.
  • Pre-cleaned, ready to use
  • Color frosted on one end, one side
  • Grounded edges, 45° corners
  • Autoclavable
  • Colors available: White, green, blue and yellow
The silane coating offers enhanced adhesion performance, specifically designed for histological and plastic sections. It proves effective for routine H&E stains, IHC, ISH, and frozen sections. Compatibility extends to inkjet and thermal transfer printers, as well as permanent markers for seamless marking.