Color Frosted Microscope Slides
Supplier: FOUR E#S INDUSTRIAL GROUP INC.
SLD245211 SLD245212 SLD245213 SLD245214
76970-072BX 23.1 CAD
76970-072 76970-074 76970-076 76970-078
Color Frosted Microscope Slides
Slides Microscope Slides
Four E's color frosted microscope slides made from premium super white glass for ultra clarity with higher light transmittance.
  • Pre-cleaned, ready to use
  • Color frosted on one end, one side
  • Grounded edges, 45° corners
  • Autoclavable
  • Colors available: White, green, blue and yellow
The colorfast opaque coating is resistant to commonly used laboratory solvents. The chromatic integrity stays intact even through routine H&E staining.