Plain Microscope Slides
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
SLD145110 SLD245110
76970-064BX 11.28 CAD
76970-064 76970-066
Plain Microscope Slides
Slides Microscope Slides
Four E's microscope slides made from high quality soda glass or super white glass.
  • Pre-cleaned, ready to use
  • Grounded edges, 45° corners
  • Autoclavable
Suitable for traditional identification labeling, these plain slides are manufactured using an advanced grinding process to ensure uniform pieces and consistency in flatness and smoothness. The polished edges and corners greatly reduce the risk of scratches and infection during use and handling of the slides.