Staticide® ESD Knit Gloves with Dipped Tips
Supplier: ACL Staticide
GLKT-2XL GLKT-L GLKT-M GLKT-S GLKT-XL
77200-754CS 231.36 CAD
77200-754 77200-756 77200-758 77200-760 77200-762
Staticide® ESD Knit Gloves with Dipped Tips
Gloves
These ESD Knit gloves are ideal for inspection and assembly.
  • Seamless knit construction
  • Nylon conductive carbon fiber blend yarn
  • Low moisture absorption
  • GLKT CAFE RGP: 10⁸ ohms
  • Polyurthane dipped tips
These micro-knit static control gloves offer superb comfort and durability, and includes micro-foam polyurethane-coated fingertips. Ideal for inspection and assembly work when static control is required.
Ordering information: Ordered by case, six packs (36 pairs of gloves) per case.
Packaging: Each pack contains six pairs.