J.T.Baker® BAKERBOND® Supported Liquid Extraction Plates
Supplier: Avantor
8182-02 8182-96 8183-04 8183-96
JT8182-02PK 310.42 CAD
JT8182-02 JT8182-96 JT8183-04 JT8183-96
J.T.Baker® BAKERBOND® Supported Liquid Extraction Plates
Protein Purification Protein Purification Reagents
J.T.Baker® BAKERBOND® supported liquid extraction (SLE) plates are an ideal and faster alternative to traditional liquid-liquid extraction methods and allow excellent recoveries of analytes from biological matrices.
  • Diatomaceous earth packing material
  • Simple protocol
  • Quicker and easier than liquid-liquid extraction
  • 96-well plate or cartridge formats offered
  • Can be used for protein removal
This sample preparation method can be used prior to chromatographic methods like LC-MS, HPLC and UHPLC, and has wide applicability for acids, bases and neutral compounds.