J.T.Baker® BAKERBOND® Protein Precipitation Plates
Supplier: Avantor
8181-596 8181-96
JT8181-596PK 893.24 CAD
JT8181-596 JT8181-96
J.T.Baker® BAKERBOND® Protein Precipitation Plates
Protein Purification Protein Purification Reagents
J.T.Baker® BAKERBOND® protein precipitation plates enable fast protein removal from biological samples.
  • Protein removal from plasma
  • Ideal prior to LC-MS applications
  • 96 well plate format for multiple samples
  • Vacuum manifold can be used (8183-VM)
  • Collection plates available
Proteins crash out of solution when the standard protocol is followed, this process removes proteins from the matrix that could interfere with LC-MS analyses and also avoids any protein precipitation during the HPLC analyses.