NOVA-TECH® 1000 Cleanroom Wipes
Supplier: High-Tech Conversions
NT1-1212 NT1-99
77007-972CS 607.29 CAD
77007-972 77008-052
NOVA-TECH® 1000 Cleanroom Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wipes are made from a hydro-entangled, non-woven material composed of 45% polyester and 55% cellulose.
  • Compatible with virtually any solvent for cleaning and wiping
  • Tough, absorbent, and abrasion-resistant
  • Low particle generation
  • Popular for use in ISO Class 6 (Class 1000) cleanrooms
  • Can be used in ISO Class 5 - 8 cleanrooms
  • Packaging provides lot-to-lot traceability
  • Double bagged packaging
  • Steam autoclavable
NOVA-TECH® 1000 cleanroom wipes are tough, abrasion-resistant, absorbent, and virtually free of particle generation. The synergistic blend of virgin polyester and cellulose is hydro-entangled into a uniform, super-tough fabric. These features give NOVA-TECH® wipes a high degree of absorbency and effectiveness during general wiping applications. Wipes made with this fabric are cleanroom laundered and processed, and double-bagged in a controlled environment.