Cobalt Blue® Chemo Prep Mats/Spill Pads
Supplier: High-Tech Conversions
CBCM-1611.15 CBCM-2216.10 CM-1611.50 CM-2216.50
77008-040CS 492.94 CAD
77008-040 77008-022 77007-962 77007-960
Cobalt Blue® Chemo Prep Mats/Spill Pads
Spill Control Materials
Cobalt Blue® Chemo Prep Mats/Spill Pads, available in validated sterile and nonsterile formats, are ideal for use in cleanrooms, barrier isolators, or glove boxes during HD compounding.
  • Absorbent, 3-ply material
  • Polyethylene and fiber blend
  • Non-porous polyethylene backing
  • Ideal for ISO class 5 cleanrooms and higher
  • Made in the USA
These chemo mats will quickly and safely remove spills before they spread. Validated sterile and nonsterile formats are available. The sterile option is validated to an SAL of 10⁻⁶ in accordance with standards andMore Product Information