ProWorks® Catch® Neon Green Nitrile Powder-Free Gloves
Supplier: High-Tech Conversions
GL-NT107NGFL GL-NT107NGFM GL-NT107NGFX GL-NT107NGFXX
77008-018CS 296.61 CAD
77008-018 77007-986 77007-966 77007-862
ProWorks® Catch® Neon Green Nitrile Powder-Free Gloves
Gloves
ProWorks® Catch® Neon green nitrile powder-free disposable gloves feature a heavy pyramid Grip® textured surface that provides an impeccable grip.
  • Industrial grade nitrile construction
  • Powder-free and latex-free
  • 9 mil thickness
  • Ambidextrous
The extra thickness makes these gloves more durable when working on 'rougher' jobs, while keeping hands safe and protected against harsh chemicals, solvents, and paints.
Certifications: Complies with FDA CFR Title 21 Indirect Food Additive Regulations, Part 177.
Packaging: Gloves are packaged 100 pieces per box.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Material Nitrile
Sterility Non sterile