VWR® Sample Bottles

Supplier: VWR International
VWRI210-000046 VWRI210-000044 VWRI210-000054 VWRI210-000048 VWRI210-000052 VWRI210-000049 VWRI210-000053 VWRI210-000050 VWRI210-000051 VWRI210-000047 VWRI210-000043 VWRI210-000045
VWRI210-000046CS 1079.02 CAD
VWRI210-000046 VWRI210-000044 VWRI210-000054 VWRI210-000048 VWRI210-000052 VWRI210-000049 VWRI210-000053 VWRI210-000050 VWRI210-000051 VWRI210-000047 VWRI210-000043 VWRI210-000045
VWR® Sample Bottles
Bottles Plastic Bottles
VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.

  • Made from durable, break-resistant 100% virgin PETG material
  • Suitable for frozen storage down to −70°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability

Bottles are arranged in shrink-wrapped trays and polybag packaging to prevent contamination. Bottles 30 ml and above feature volume graduations. Non sterile bottles are autoclavable.

Certifications: US FDA 21CRF, USP Class VI compliant resin.

Accessories information: Spare closures are available for specific application requirements.

Packaging: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR