Bronze Latex Powder-Free Examination Gloves
Supplier: High-Tech Conversions
BRZ642 BRZ645 BRZ646 BRZ648
77008-044CS 156.8 CAD
77008-044 77007-956 77007-870 77008-004
Bronze Latex Powder-Free Examination Gloves
Gloves
Bronze Latex Powder-Free Exam Gloves are a quality choice at an economical and affordable cost.
  • Single-use only
  • Easy donning and doffing
Though these gloves are made in lighter weight and thinner gauge, they are strong enough for normal everyday examination use in dental, medical, and laboratory applications.
Packaging: Gloves are packaged 100 pieces per box (90 pieces per box for size XL).