SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W832018 W832022 W832024
89044-832EA 33.85 CAD
89044-832 89044-836 89044-838
SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Bottles Plastic Bottles
Natural HDPE bottles with screw cap.
Perfectly adapted with Calibrex™ bottle top dispensers. Corresponding neck adapters supplied with the dispensers.