All Weather Permanent Adhesive Vinyl Labels
Supplier: Brady Worldwide
176849 176850 176851
77282-348RL 147.19 CAD
77282-348 77282-350 77282-352
All Weather Permanent Adhesive Vinyl Labels
Labels
Get long-lasting durability for indoor/outdoor asset identification with these pre-size vinyl (B-595) labels. Engineered to withstand extreme temperatures.
  • Permanent acrylic adhesive
  • Gloss finish
  • Excellent outdoor durability, cold-temperature application
  • Abrasion-resistant, chemical-resistant, and oil-resistant
  • Service temperature: −40 to180 °F
Extra-durable labels resist abrasions, chemicals, and oils. Labels offer an 8-10 year average outdoor durability in temperatures ranging from −40 to 180 °F (−40 to 82 °C). They can be applied as low as 0 °F (−18 °C). High-gloss vinyl labels feature a permanent acrylic adhesive and are thermal transfer printable. Available in white, yellow, and orange for multiple applications.
For use with M710 printers. Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material; use part number M7-R10000.