Solvent and Chemical Resistant Polyester Labels
Supplier: Brady Worldwide
176998 176999 177000 177001 177002
77282-378RL 355.56 CAD
77282-378 77282-380 77282-382 77282-384 77282-386
Solvent and Chemical Resistant Polyester Labels
Labels
High-performance polyester (B-488) withstands numerous solvents and variable temperatures when applied to various surfaces, including stainless steel and polypropylene.
  • Permanent acrylic adhesive
  • Matte finish
  • Autoclavable, cryogenic storage material
  • Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, and weather-resistant
  • UL approved
Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable. Ideal for general and barcode labeling on flat, smooth surfaces. Service temperature of −40 to 160 °C (−40 to 320°F).
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material; use part number B30-R4300.