High-Adhesion Self-Laminating Wire and Cable Labels
Supplier: Brady Worldwide
170984 170985 170986 170987 170988 170989 170990
77282-256RL 916.19 CAD
77282-256 77282-258 77282-260 77282-262 77282-264 77282-266 77282-268
High-Adhesion Self-Laminating Wire and Cable Labels
Use high-adhesive, self-laminating labels for tough conditions like high-humidity, small diameter wires, and newer wire jacketing materials. Bulk label rolls are ideal for continuous printing, reducing the frequency of label changeouts. Wraptor™ A6200 bulk label roll adapter required to utilize bulk label rolls.
  • Permanent acrylic adhesive
  • Matte finish
  • Self-laminating, self-extinguishing material
  • Abrasion-resistant, chemical-resistant, humidity-resistant, oil-resistant, water-resistant, and weather-resistant
  • Service temperature: −40 to 158 °F
Dependable, long-term adhesion to small diameter wires. Labels will stay adhered in even the most humid environments. Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC. Labels feature a white printable area and a clear overlaminating area that protects the print. Labels are self-extinguishing and resist chemicals, humidity, oil, and water.
Use with Wraptor™ A6200 bulk label roll adapter and Wraptor™ A6200 wrap printer applicator and Wraptor™ A6200 bulk label roll adapter (required). A ribbon is required to print this material; use part number A62-R4300.