Self-laminating Vinyl Wrap-Around Labels
Supplier: Brady Worldwide
170977 170978 170979 170980 170981 170982 170983
77282-242RL 933.65 CAD
77282-242 77282-244 77282-246 77282-248 77282-250 77282-252 77282-254
Self-laminating Vinyl Wrap-Around Labels
Labels
Use high-performance, self-laminating labels to identify wires and cables. Bulk label rolls are ideal for continuous printing, reducing the frequency of label changeouts. Wraptor™ A6200 Bulk label roll adapter required to utilize bulk label rolls.
  • Permanent acrylic adhesive
  • Matte finish
  • Flame-retardant, self-laminating material
  • Chemical-resistant, humidity-resistant, oil-resistant, water-resistant, and weather-resistant
  • Service temperature: −94 to 158 °F
  • UL approved
Protect printed text using a self-laminating format. Labels feature a white printable zone with translucent overlaminating area. Labels are self-extinguishing and resist chemicals, humidity, oil, and water. Highly conformable with a durable, permanent acrylic adhesive for indoor/outdoor use.
Use with Wraptor™ A6200 Wrap printer applicator and Wraptor™ A6200 bulk label roll adapter (required). A ribbon is required to print this material; use part number R4300.