Clear Acrylic Overlaminate for BradyJet™ Labels
Supplier: Brady Worldwide
175440 175441 175442 175443 175444
77282-290RL 273.92 CAD
77282-290 77282-292 77282-294 77282-296 77282-298
Clear Acrylic Overlaminate for BradyJet™ Labels
Labels
Achieve up to 10 years of outdoor durability through improved UV and abrasion resistance with acrylic overlaminate for BradyJet™ labels.
  • Permanent acrylic adhesive
  • Gloss finish
  • Optically clear material
  • Service temperature: −20 to 180 °F
No need to cut to size, simply peel and apply with these pre-sized rolls that are perfectly sized to their corresponding BradyJet™ labels. Overlaminate is a quarter inch wider than its corresponding BradyJet™ label to ensure complete coverage.