Catalepsy Bar Test
Supplier: MAZE ENGINEERS
5708 5709
76670-300EA 10522.8 CAD
76670-300 76670-302
Catalepsy Bar Test
Animal Supplies Animal Environmental Enrichment
The Catalepsy Bar test is an alternative to the Rotarod apparatus, and was developed to test motor coordination and motor impairments.
  • Lane width: 10 cm
  • Bar diameter: 0.7 cm
  • Distance of the bar above the floor is adjustable between 3 to 8 cm (6 cm average)
  • Separators are made of black acrylic
The Catalepsy Bar Test involves placing the forepaws of a rodent onto a bar and timing how long it takes the subject to remove one or both paws from the bar. The automated Catalepsy Bar test has 4 or 6 lanes for the simultaneous study of multiple subjects with adjustable bar height to ensure perfect placement for each subject. Sensors to detect when an animal removes its paws from the bar and included software logs the time at which this occurs. Models available for mice and rats.