Learned Helplessness Apparatus
Supplier: MAZE ENGINEERS
5401 5402
76670-274EA 8216 CAD
76670-274 76670-276
Learned Helplessness Apparatus
Animal Supplies Animal Environmental Enrichment
The learned helplessness paradigm is a widely used animal model of depression, displaying exposure to inescapable shock resulting in dramatic deficits in emotional expression, associative learning, and behavioral coping when presented to an aversive but escapable stimulus.
  • Includes contexual plates for different colors and patterns and shock plates for D/C shock delivery
The learned helplessness paradigm is a widely used animal model of depression originally described by Overmier and Seligman (1967), who observed that exposure to inescapable shockMore Product Information