Automated Radial Arm Maze
Supplier: MAZE ENGINEERS
1901 1902
76669-836EA 25016.68 CAD
76669-836 76669-838
Automated Radial Arm Maze
Animal Supplies Animal Environmental Enrichment
The automated 8 arm Radial Mazes creates an entirely automated environment with automated doors, reward delivery, and video tracking integration.
  • Quiet automated pnaeumatic doors minimize human intervention
  • Easy to clean acrylic
  • Matte, non-reflective surfaces
  • Odor-free
The automated 8 Arm Radial maze is an apparatus widely validated for spatial learning tasks using external cues and arm baiting. Automated doors, reward delivery, and video tracking integration minimize anxiety and fear created by the human experimentor while collecting high volumes of data.
Ordering information: The Automated Radial Arm Maze requires software to function (see accessory options). Software packages are designed for use with the Automated Radial Maze and video tracking software such as EthoVision or ANY-Maze.