Successive Alleys Tests
Supplier: MAZE ENGINEERS
5109 5110
76670-252EA 4187.03 CAD
76670-252 76670-254
Successive Alleys Tests
Animal Supplies Animal Environmental Enrichment
The Successive Alleys test is a novel anxiety test that utilizes fear of open spaces as an assay.
  • Acrylic construction
  • Easy to assemble
  • Easy storage, cleaning, assembly, and disassembly
In the Successive Alleys test, the most distal regions are white, open, and narrow, which creates an anxiogenic environment. The most proximal, dark areas create anxiogenic areas that encourage natural aversion of open spaces.
The Successive Alleys test for mouse features 4 alleys. Alley 1 is black, and measures 8.5W×25H cm. Alley 2 is grey and measures 8.5W×5H cm.More Product Information