Gait Tests
Supplier: MAZE ENGINEERS
5008 5009
76670-196EA 3981.6 CAD
76670-196 76670-198
Gait Tests
Animal Supplies Animal Environmental Enrichment
The Gait test is a useful apparatus to measure motor lesions and other dysfunctions related to gait.
  • Simple and reliable
  • Easy to clean
In the Gait test, the forepaw of the rodent is wetted with blue black ink (included) and placed on one end of the runway, which is covered with a strip of paper. The stride length of forelimbs are measured manually as the distance between two forepaw prints. This apparatus comes with an optional lighting device to encourage motivation to the dark housing area. Sizing for mice and rats available.