Horizontal and Vertical Grid Test
Supplier: MAZE ENGINEERS
5003 5004 5001 5002
76670-186EA 1242.94 CAD
76670-186 76670-188 76670-182 76670-184
Horizontal and Vertical Grid Test
Animal Supplies Animal Environmental Enrichment
The horizontal grid apparatus allows for the study of neuromuscular strength and coordination in rodents. The vertical grid test is similar to the horizontal grid test to assess motor function, neuromuscular strength, and coordination.
  • Acrylic frame construction with metal mesh
  • Simple to use and easy to clean, assemble, and disassemble
The horizontal grid apparatus is a horizontally positioned grid box mounted with plexiglass, which allows the subect to grab onto the grids as it climbs across while upside down. The vertical grid apparatus is a vertically positioned grid box mounted with acrylic, and this allows the subject to grab on the grids as it climbs down.