Visual Burrow System
Supplier: MAZE ENGINEERS
4909 4910
76670-158EA 6557.03 CAD
76670-158 76670-160
Visual Burrow System
Animal Supplies Animal Environmental Enrichment
The Visual Burrow System (VBS) was developed by Bob Blanchard and Caroline Blanchard in the late 1980s. The VBS is commonly used in the evaluation of social behaviors.
  • Default black acrylic construction
  • Easy to clean
The Visual Burrow System (VBS) mimics the natural environment of rodents, enabling observation of natural social interactions that would have been otherwise impossible to observe in the wild. Each colony in the Visual Burrow System is housed in a rectangular acrylic chamber. Three chambers are positioned behind a barrier wall in theMore Product Information