Resident Intruder Tests
Supplier: MAZE ENGINEERS
4903 4904
76670-146EA 4977.03 CAD
76670-146 76670-148
Resident Intruder Tests
Animal Supplies Animal Environmental Enrichment
The Resident Intruder test is a chamber used for social defeat paradigms.
  • Clear and white acrylic chamber
  • Metal mesh top
  • Easy to clean
Composed of a clear cage front for video recording, opaque sides and back, wire top, and a bottom lip to keep in bedding. Easy to clean by spraying and wiping with 70% ethanol, without losing clarity of front wall.