Lashley III Maze
Supplier: MAZE ENGINEERS
4705 4706
76670-094EA 3981.6 CAD
76670-094 76670-096
Lashley III Maze
Animal Supplies Animal Environmental Enrichment
The Lashley III maze is a test used for learning and memory. It utilizes a low stress environment and looks into route learning of a maze under repeated trials.
  • Acrylic construction
  • Non-reflective surfaces
In typical protocols, no stress, food deprivation, or other stimuli are used. A start box, a maze body, and a goal box are contained in the complete apparatus. The goal box can be filled with bedding similar to a home cage to reward the subjects, and to motivate them toward completion of the task. A large red base is not included in the order, butMore Product Information