Forced Swim Test
Supplier: MAZE ENGINEERS
3701 3702 3703
76669-970EA 1874.94 CAD
76669-970 76669-972 76669-974
Forced Swim Test
Animal Supplies Animal Environmental Enrichment
The Forced Swim Test (FST) is commonly used to assess anti-depressant-like behavior in rodents.
  • Acrylic construction
  • Easy to clean
The Forced Swim Tests involves the scoring of active movements such as swimming and climbing, versus passive immobile behavior while swimming in a cylinder.