Elevated Plus Maze
Supplier: MAZE ENGINEERS
3301 3302
76669-910EA 3981.6 CAD
76669-910 76669-912
Elevated Plus Maze
Animal Supplies Animal Environmental Enrichment
The Elevated Plus Maze and accessories measure anxiety-like behavior in rodents.
  • All open arms include 1 cm high-end plates
  • Acrylic construction
  • Easy to assemble and disassemble
The Elevated Plus Maze measures anxiety-like behavior. The test is based on the natural aversion of rodents to open and elevated areas, as well as on their natural spontaneous exploratory behavior in novel environments. This test is commonly used with pharmacotherapeutic validation and screening for anti-anxiogenic and anti-anxiogenic properties.
Ordering information: The optional Raised edge lip for the Elevated Plus Maze (mouse or rat) is available with 0.5 cm or 1 cm height. Please specify the required height when ordering.