Open Field
Supplier: MAZE ENGINEERS
3203 3204 3205 3206 3221 3222 3201 3202
76669-886EA 1664.28 CAD
76669-886 76669-888 76669-890 76669-892 76669-904 76669-906 76669-882 76669-884
Open Field
Animal Supplies Animal Cages
The Open Field and accessories are used to study Thigotaxis-related behaviors in rodents.
  • Acrylic construction with matte finish for non-shine
  • Easy assembly and disssembly with a removable base
  • Easy to clean
Thigmotaxis in the open field is used to evaluate anxiolytic, anxiogenic, and non-pharmacological treatments as well as genetic manipulations. Ambulation is the most common behavior studied using the open field, but others such as latency or rearing can also be measured. In addition, objects can be added for a modification similar to the novel object recognition field.