Automated Repeated Acquisition and Performance Chambers
Supplier: MAZE ENGINEERS
6304 6303
76670-366EA 18938.94 CAD
76670-366 76670-364
Automated Repeated Acquisition and Performance Chambers
Animal Supplies Animal Environmental Enrichment
The repeated acquisition and performance chamber (RAPC) is an acrylic device with a start and end goal box. Correct entry to the final destination/goal box allows access to a reward determined by the experimenter.
  • Acrylic construction
The interior main body consists of five compartments with four transparent panels. One-way doors separate each of the compartments and allow access to successive compartments. Correct entry to the final destination or goal box allows access to a reward determined by the experimenter.
Ordering information: SoftwareMore Product Information