Automated Hole Boards
Supplier: MAZE ENGINEERS
4503 4504
76670-054EA 7352.28 CAD
76670-054 76670-056
Automated Hole Boards
Animal Supplies Animal Environmental Enrichment
The automated hole board system allows for easy detection of nose poke behavior in the context of an open field environment.
  • Includes 16, 9, and 4 hole boards and open field
  • 16 nose poke detection cups included
  • Clear wall set included
  • Includes automated pneumatic downward facing doors
Hole boards included in the order are 16, 9, and 4 holes. The apparatus provided allows for maximum flexibility, including the ability to interchange acrylic walls for colored walls, nose pokes, and floor interfaces. Each order comes with the conductor software to allow for easy control of the hole board apparatus as well as logging of data during automated experiments. Software is also available separately as an accessory option.