Parallel Rod Floor Tests
Supplier: MAZE ENGINEERS
6501 6502 6503 6504
76670-376EA 1664.28 CAD
76670-376 76670-378 76670-380 76670-382
Parallel Rod Floor Tests
Animal Supplies Animal Environmental Enrichment
The parallel rod floor test was originally used as a simple model of ataxia in rodents.
  • Clear acrylic and metal grid
  • Easy to clean, assemble, and disassemble
The parallel rod floor test was originally described in the literature by Kamans and Crabbe (2007) as a simple model of ataxia in mice. The test allows researchers to quantify differences in motor coordination.