Social Reward Chambers
Supplier: MAZE ENGINEERS
4907 4908
76670-154EA 2506.94 CAD
76670-154 76670-156
Social Reward Chambers
Animal Supplies Animal Environmental Enrichment
The social reward chamber is designed to understand the effect of social rewards on the social preferences of the subject.
  • Clear acrylic construction
Unlike the existing models that are exclusively based on the Conditioned Place Preference (CPP) paradigm and require memory association (Trezza et al., 2011), the Social Reward Chamber is based on Operant Social Preference (OSP) tasks that take advantage of visual stimulus (Borland et al., 2017).
The social reward chamber for mouse features a main chamber measuring 33×22×20 cm, and two additional chambers measuring 17×12×20 cm. The social reward chamber for rat features a main chamber measuring 50×33×30 cm, and two additional chambers measuring 25×18×30 cm.