Elevated Zero Mazes
Supplier: MAZE ENGINEERS
4001 4002
76669-998EA 3981.6 CAD
76669-998 76670-000
Elevated Zero Mazes
Animal Supplies Animal Environmental Enrichment
The elevated zero maze is an elevated, ring-shaped runway with the same amount of area devoted to adjacent open and closed quadrants, similar to the elevated plus maze to study anxiety behaviors.
  • Acrylic construction
  • Odor-free
  • Matted finish for non-reflective surfaces
  • Easy to clean
  • Easy assembly and disssembly with a removable base
The elevated zero maze is very similar to the elevated plus maze, but with the center region of the elevated plus maze removed. It has been pharmacologically validated with various anxiolytic drugs.
The design is created with an elevated circular platform with two walled, enclosed portions as well as two open portions, allowing for clear interpretations of murine behavior. Because mice tend to avoid open-spaced areas, the animals will preferentially spend more time in the enclosed walled area.