Automated Empathy Assays

Supplier: MAZE ENGINEERS
7301 7302
76670-392EA 12618.94 CAD
76670-392 76670-394
Automated Empathy Assays
Animal Supplies Animal Environmental Enrichment
The empathy assay is used to interrogate observational fear. Emotional empathy can be studied as a precursor to a fear response, arising from observing a subject experiencing an aversive stimulus.

  • Speaker frequency: 100 to 40000 Hz
  • Speaker intensity: 1 to 150 dB
  • Replaceable acrylic plates with multiple contexts allow for contextual learning experiments
  • Default plate patterns: gray, white, black, chessboard, vertical stripes

The novel environment tends to reduce the effect of the previous day’s fear conditioning during the 24 hours memory test. This apparatus comes with two key components; a double-chambered acrylic chamber with two-foot, shock-controlled grids, each of which can be controlled independently with the included conductor software. This apparatus is then placed inside of a standard, basic isolation chamber. Upgrades in this chamber include sound and light cues (please inquire for more information). The apparatus for the Empathy assay includes a sound-attenuating cubicle with a fan within which an observational fear-conditioning chamber is placed. The fear-conditioning chamber is composed of two chambers that are separated by a transparent Plexiglas divider. The chambers feature stainless steel rods placed parallel on the floor. A programmable, computer-controlled shocking system is also used to administer foot shocks. Two plates are included and both plates can be controlled separately.

Ordering information: Software package (see accessory option) is designed for use with the automated empathy assay as stand alone software.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR