Fear Conditioning Chambers
Supplier: MAZE ENGINEERS
5801 5802 5803 5804 5805 5806
76670-304EA 12429.34 CAD
76670-304 76670-306 76670-308 76670-310 76670-312 76670-314
Fear Conditioning Chambers
The fear conditioning chamber is a behavioral task widely used in neuroscience to assess associative learning.
  • Includes one enclosure, speaker, lights, shocker, and sound attenuation chamber
The fear conditioning chamber is a carefully designed chamber designed to allow new scientific avenues, and to allow easy data collection and experimental execution. The chamber comes with an isolation chamber with speakers, dB detector, dual IR/visible light generation. The contexual cage features easy to replace acrylic plates and smooth shock delivery.
Ordering information: The optional accessory software package is designed for use with the fear conditioning chamber and video tracking software such as EthoVision or ANY-Maze.