Drosophila Shallow Chambers
Supplier: MAZE ENGINEERS
8506 8507
76670-418EA 4450.35 CAD
76670-418 76670-420
Drosophila Shallow Chambers
Animal Supplies Animal Environmental Enrichment
The shallow drosophila chamber was first described by Simon et al (2010) as an innovative design to restrict flies to a shallow volume of space, forcing all behavioral interactions to take place within a monolayer of individuals.
  • 7 cm model is available for courtship and aggression studies
  • 13 cm model is available for large groups of drosophila
  • 11 degree sloped flooring, glass ceiling, LED multi-light display
  • Visual stimulus of a 12×12" array of 850 nm IR LED mounted underneath the chambers for backlighting
The chamber design minimizes the frequency that flies occlude or obscure each other, and promotes a greater number of flies to move throughout the center of the chamber. This increases the frequency of interactions and improves the quality of data collected by digital video.