Hoarding Apparatus
Supplier: MAZE ENGINEERS
4913 4914
76670-162EA 5767.02 CAD
76670-162 76670-164
Hoarding Apparatus
Animal Supplies Animal Environmental Enrichment
The Hoarding apparatus is used to study a species-typical behavior manifested by transferring food to a secure place for later consumption.
  • Clear acrylic construction
  • Easy to clean
  • Simple and easy to use
The Hoarding apparatus allows for behavioral study using eight rows of connecting home chambers, each connected to a hoarding tube.