Self Administration Runways
Supplier: MAZE ENGINEERS
4707 4708
76670-098EA 5240.34 CAD
76670-098 76670-100
Self Administration Runways
Animal Supplies Animal Environmental Enrichment
The self-administration runway is made of acrylic and is composed of three main parts; the start box, the goal box, and the runway. This apparatus is typically used in research for drug self administration.
  • Acrylic construction
  • Non reflective surfaces
Drugs, alcohol, and other substances can be placed at the goal box. The apparatus has different sizes for mice and rats, and is available in multiple colors for a start box/end box and three runways with small barriers in the floor.