Hebb-Williams Mazes
Supplier: MAZE ENGINEERS
4701 4702
76670-086EA 6557.03 CAD
76670-086 76670-088
Hebb-Williams Mazes
Animal Supplies Animal Environmental Enrichment
The Hebb-Williams Maze (HWM) is a type of behavioral test used to assess spatial learning and memory in rodents.
  • Acrylic construction
  • Non reflective surfaces
The HWM consists of a central platform with several arms radiating out from it. The maze is made of acrylic and the arms can be baited with food or other rewards. The test is typically conducted by placing the animal on the central platform and allowing it to explore the maze. The animal’s movements are then tracked, and different measures such as the number of arm entries, the time spent in each arm, and the number of errors made can be used to assess the animal’s spatial learning and memory abilities.