Tail Suspension Tests
Supplier: MAZE ENGINEERS
4201 4202 4204
76670-014EA 3360.14 CAD
76670-014 76670-016 76670-018
Tail Suspension Tests
Animal Supplies Animal Environmental Enrichment
The tail suspension test is a standard test of depression in rodents.
  • Acrylic construction
  • Non reflective surface
  • Easy customization
Since its creation in 1985, the tail suspension test is a standard test of depression (Steru et al., 1985). The standard protocol is to hang mice to the apparatus by their tail.