Barnes Maze
Supplier: MAZE ENGINEERS
3605 3610 3611 3612 3613 3641 3642 3601 3602
76669-950EA 7083.68 CAD
76669-950 76669-958 76669-960 76669-962 76669-964 76669-966 76669-968 76669-942 76669-944
Barnes Maze
Animal Supplies Animal Environmental Enrichment
The Barnes Maze is used to assess spatial learning and memory impairments in rodent models.
  • Includes one target box with the option to add more holes
  • Entire top and dark escape box rotate for customization
  • Easy to clean with a removable escape box holder
  • False floor modification available for added versatility
  • Thick acrylic top eliminates visual cues
The Barnes Maze is often used to assess spatial learning and memory impairments in rodent models of neurological disorders, including Alzheimer’s disease and traumatic brain injury. The task is repeated over several trials, and researchers measure performance parameters such as the time taken to find the escape box, the number of errors made (visiting incorrect holes), and the overall strategy employed by the animal to solve the task.
For Barnes Maze modifications, the delayed matching to place Barnes Maze is a modification of the Barnes Maze using a well patterned Barnes Maze with 16 holes on the outer ring, 16 on the middle ring, and 8 holes on the inner ring with distance of 50, 35, and 20 cm to the center of platform, respectively. The randomized Barnes Maze increases the task difficulty of the classic Barnes Maze, leading to a decrease in the serial strategy of goal hole choice. The cheeseboard maze is a circular maze with 32 wells forming a radial pattern. The wells were spaced radially resulting in a similar shape to a radial arm maze with wells distributed throughout the table.