Bumper Guards for Volumetric Flasks
Supplier: United Scientific Supplies
BMP100PK5 BMP250PK5 BMP25PK5 UNBMP1000 UNBMP500
470353-220PK 36.66 CAD
470353-220 470353-246 470353-224 470353-296 470353-260
Bumper Guards for Volumetric Flasks
Supports Flask Supports
Protect volumetric flasks from accidentally being tipped over.
  • Wide design to cushion flasks if tipped over
  • Made of high-quality silicone
Silicone bumpers slip easily onto any volumetric flask. In the event of a flask being accidentally tipped over, the wide bumpers cushion the neck from hitting the table surface. Color-coded and available in five different sizes.