VWR® EZLabpure™ Round Carboys, with 83B VersaCap®
Supplier: VWR International
76461-398 76461-400 76461-402 76461-404 152-B191-FLS 152-C191-FLS 155-E191-FLS
76461-398EA 137.04 CAD
76461-398 76461-400 76461-402 76461-404 77296-500 77296-502 77296-504
VWR® EZLabpure™ Round Carboys, with 83B VersaCap®
Bottles Carboys
VWR® EZLabpure™ round carboys are manufactured using high-purity virgin USP Class VI medical grade polypropylene.
  • Carboys can be used for storing and shipping reagents, chemicals, culture media, and standards
  • Polypropylene carboys are autoclavable at 121 °C for twenty minutes
  • Can be used at a temperature below 0 °C
  • Superior chemical resistance
A premium line of high-quality, round HDPE and polypropylene (PP) laboratory carboys with the patented VersaCap® technology. These durable, rugged, and shatterproof round carboys are a great alternative to glassMore Product Information