Anti-CNPase Mouse Monoclonal Antibody [clone: 1H10]

Supplier: ANTIBODIES.COM LLC
A85413-50 A85413-100
77068-130EA 676.4 CAD
77068-130 77080-484
Anti-CNPase Mouse Monoclonal Antibody [clone: 1H10]
Antibodies
Mouse monoclonal (1H10) antibody to CNPase for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1000, ICC/IF: 1:50

Type: Primary
Antigen: CNPase
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR