Anti-CNPase Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85412-100 A85412-50
77072-266EA 877.69 CAD
77072-266 77077-040
Anti-CNPase Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CNPase for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:1000 to 1:200

Type: Primary
Antigen: CNPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR