Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 3G12]

Supplier: ANTIBODIES.COM LLC
A85361-100 A85361-50
77065-770EA 877.69 CAD
77065-770 77072-518
Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 3G12]
Antibodies
Mouse monoclonal (3G12) antibody to 14-3-3 eta for WB, ICC/IF and IHC with samples derived from Human, Bovine, Porcine, Canine, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:5000, ICC/IF: 1:1000, IHC: 1:100

Type: Primary
Antigen: 14-3-3 eta
Clonality: Monoclonal
Clone: 3G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Bovine;Porcine;Dog;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR